Assay Details
| Gene Symbol: LOC769174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079740.1 | NP_001073208.1 | 6 - 7 | 630 | 107 | |
| GenBank mRNA | DQ241793.1 | - | 6 - 7 | 630 | 107 | |
More Information
Set Membership: |
|

LOC769174
Genomic Map