Assay Details
| Gene Symbol: STARD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079742.1 | NP_001073210.1 | 4 - 5 | 375 | 70 | |
| XM_015280436.1 | XP_015135922.1 | 2 - 3 | 198 | 70 | ||
| GenBank mRNA | AJ720217.1 | - | 4 - 5 | 375 | 70 | |
More Information
Set Membership: |
|

STARD4
Genomic Map