Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079751.2 | NP_001073219.1 | 4 - 5 | 484 | 56 | |
| XM_015299946.2 | XP_015155432.1 | 3 - 4 | 357 | 56 | ||
| XM_015299947.2 | XP_015155433.1 | 3 - 4 | 483 | 56 | ||
| XM_015299948.2 | XP_015155434.2 | 4 - 5 | 495 | 56 | ||
| GenBank mRNA | AJ719606.1 | - | 4 - 5 | 485 | 56 | |
More Information
Set Membership: |
|

MBD3
Genomic Map