Assay Details
| Gene Symbol: RRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079759.2 | NP_001073227.1 | 3 - 4 | 498 | 69 | |
| GenBank mRNA | AY339626.1 | - | 3 - 4 | 502 | 69 | |
More Information
Set Membership: |
|

RRH
Genomic Map