Assay Details
| Gene Symbol: PFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079760.1 | NP_001073228.1 | 2 - 3 | 371 | 73 | |
| XM_025153418.1 | XP_025009186.1 | 2 - 3 | 329 | 73 | ||
| GenBank mRNA | AJ719884.1 | - | 2 - 3 | 371 | 73 | |
More Information
Set Membership: |
|

PFN2
Genomic Map