Assay Details
Gene Symbol: BRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079761.1 | NP_001073229.1 | 4 - 5 | 636 | 69 | |
XM_015275345.1 | XP_015130831.1 | 3 - 4 | 429 | 69 | ||
GenBank mRNA | AJ719925.1 | - | 4 - 5 | 636 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
