Assay Details
| Gene Symbol: CHEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080107.1 | NP_001073576.1 | 1 - 2 | 313 | 65 | |
| XM_015275342.2 | XP_015130828.1 | 1 - 2 | 285 | 65 | ||
| GenBank mRNA | EF118300.1 | - | 1 - 2 | 313 | 65 | |
More Information
Set Membership: |
|

CHEK2
Genomic Map