Assay Details
| Gene Symbol: BDKRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080720.1 | NP_001074189.1 | 1 | 849 | 117 | |
| XM_015287568.2 | XP_015143054.1 | 2 | 1333 | 117 | ||
More Information
Set Membership: |
|

BDKRB1
Genomic Map