Assay Details
| Gene Symbol: GOSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080864.2 | NP_001074333.2 | 3 - 4 | 262 | 85 | |
| XM_015299229.2 | XP_015154715.1 | 3 - 4 | 270 | 85 | ||
| XM_025143972.1 | XP_024999740.1 | 3 - 4 | 271 | 85 | ||
| GenBank mRNA | AJ447074.1 | - | 3 - 4 | 240 | 85 | |
| AJ719943.1 | - | 3 - 4 | 246 | 85 | ||
| CB016763.1 | - | 3 - 4 | 216 | 85 | ||
| CN235529.1 | - | 3 - 4 | 262 | 85 | ||
More Information
Set Membership: |
|

GOSR2
Genomic Map