Assay Details
| Gene Symbol: SRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080865.1 | NP_001074334.1 | 1 - 2 | 77 | 69 | |
| GenBank mRNA | AJ719556.1 | - | 1 - 2 | 77 | 69 | |
More Information
Set Membership: |
|

SRI
Genomic Map