Assay Details
| Gene Symbol: SALL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080872.1 | NP_001074341.1 | 3 - 4 | 3017 | 74 | |
| XM_015296420.2 | XP_015151906.1 | 3 - 4 | 2991 | 74 | ||
| XM_015296421.2 | XP_015151907.1 | 3 - 4 | 2933 | 74 | ||
| XM_025142271.1 | XP_024998039.1 | 3 - 4 | 2922 | 74 | ||
| GenBank mRNA | AJ851732.1 | - | 3 - 4 | 3017 | 74 | |
| AY342354.1 | - | 3 - 4 | 3008 | 74 | ||
| CR391189.1 | - | 1 - 2 | 175 | 74 | ||
More Information
Set Membership: |
|

SALL4
Genomic Map