Assay Details
| Gene Symbol: PRLH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082419.1 | NP_001075888.1 | 2 - 3 | 262 | 61 | |
| GenBank mRNA | EF418015.1 | - | 2 - 3 | 262 | 61 | |
More Information
Set Membership: |
|

PRLH
Genomic Map