Assay Details
| Gene Symbol: CXCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083362.1 | NP_001076831.1 | 1 | 1048 | 83 | |
| XM_015289178.2 | XP_015144664.1 | 2 | 1159 | 83 | ||
| XM_025152261.1 | XP_025008029.1 | 2 | 2210 | 83 | ||
| GenBank mRNA | AY332746.1 | - | 1 | 1048 | 83 | |
More Information
Set Membership: |
|

CXCR7
Genomic Map