Assay Details
Gene Symbol: DOK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083363.1 | NP_001076832.1 | 2 - 3 | 349 | 63 | |
XM_015293824.2 | XP_015149310.2 | 2 - 3 | 431 | 63 | ||
XM_025154874.1 | XP_025010642.1 | 2 - 3 | 292 | 63 | ||
GenBank mRNA | EF051736.1 | - | 2 - 3 | 349 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
