Assay Details
| Gene Symbol: SERPINE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083920.1 | NP_001077389.1 | 9 - 10 | 1271 | 112 | |
| XM_015276895.2 | XP_015132381.1 | 9 - 10 | 1369 | 112 | ||
| XM_015276896.2 | XP_015132382.1 | 8 - 9 | 1351 | 112 | ||
| XM_015276897.2 | XP_015132383.1 | 8 - 9 | 1170 | 112 | ||
| XM_015276898.2 | XP_015132384.1 | 7 - 8 | 1175 | 112 | ||
| XM_025153343.1 | XP_025009111.1 | 8 - 9 | 1218 | 112 | ||
| XM_025153344.1 | XP_025009112.1 | 7 - 8 | 1217 | 112 | ||
| GenBank mRNA | BX934893.1 | - | 9 - 10 | 1271 | 112 | |
| CR352494.1 | - | 7 - 8 | 1190 | 112 | ||
| CV860845.1 | - | 5 - 6 | 812 | 112 | ||
More Information
Set Membership: |
|

SERPINE2
Genomic Map