Assay Details
| Gene Symbol: DUSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085359.2 | NP_001078828.1 | 3 - 4 | 828 | 77 | |
| GenBank mRNA | AF026522.1 | - | 3 - 4 | 695 | 77 | |
| CR387287.1 | - | 3 - 4 | 614 | 77 | ||
More Information
Set Membership: |
|

DUSP1
Genomic Map