Assay Details
| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097531.1 | NP_001091000.1 | 3 - 4 | 342 | 103 | |
| XM_025147346.1 | XP_025003114.1 | 3 - 4 | 350 | 103 | ||
| XM_025147347.1 | XP_025003115.1 | 3 - 4 | 328 | 103 | ||
| GenBank mRNA | AJ719359.1 | - | 3 - 4 | 342 | 103 | |
More Information
Set Membership: |
|

ZNF706
Genomic Map