Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097532.3 | NP_001091001.1 | 1 - 2 | 205 | 115 | |
| GenBank mRNA | AJ720510.1 | - | 1 - 2 | 161 | 115 | |
| BX930878.2 | - | 1 - 2 | 107 | 115 | ||
| BX932133.2 | - | 1 - 2 | 153 | 115 | ||
More Information
Set Membership: |
|

ATP6V0E1
Genomic Map