Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097536.1 | NP_001091005.1 | 2 - 3 | 226 | 70 | |
| GenBank mRNA | AJ721047.1 | - | 2 - 3 | 226 | 70 | |
More Information
Set Membership: |
|

SRP9
Genomic Map