Assay Details
| Gene Symbol: TBPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098853.1 | NP_001092323.1 | 6 - 7 | 906 | 67 | |
| XM_015287569.2 | XP_015143055.1 | 7 - 8 | 4027 | 67 | ||
| XM_015287570.2 | XP_015143056.1 | 6 - 7 | 3859 | 67 | ||
More Information
Set Membership: |
|

TBPL2
Genomic Map