Assay Details
| Gene Symbol: TAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099357.1 | NP_001092827.1 | 6 - 7 | 1632 | 65 | |
| XM_015294990.2 | XP_015150476.1 | 7 - 8 | 1814 | 65 | ||
| XM_015294991.2 | XP_015150477.1 | 7 - 8 | 1767 | 65 | ||
| GenBank mRNA | AJ843262.1 | - | 6 - 7 | 1632 | 65 | |
| CR388508.1 | - | 1 - 2 | 79 | 65 | ||
| CR390666.1 | - | 5 - 6 | 1036 | 65 | ||
| JF794484.1 | - | 6 - 7 | 1632 | 65 | ||
| JF794485.1 | - | 6 - 7 | 1632 | 65 | ||
| JF794486.1 | - | 6 - 7 | 1632 | 65 | ||
| JF794487.1 | - | 6 - 7 | 1632 | 65 | ||
| JF794488.1 | - | 6 - 7 | 1632 | 65 | ||
| JF794489.1 | - | 6 - 7 | 1632 | 65 | ||
| JF794490.1 | - | 6 - 7 | 1632 | 65 | ||
More Information
Set Membership: |
|

TAP2
Genomic Map