Assay Details
| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100287.1 | NP_001093757.1 | 8 - 9 | 1516 | 69 | |
| XM_025147291.1 | XP_025003059.1 | 9 - 10 | 1624 | 69 | ||
| XM_025147292.1 | XP_025003060.1 | 8 - 9 | 1513 | 69 | ||
| XM_025147293.1 | XP_025003061.1 | 10 - 11 | 1715 | 69 | ||
| XM_025147294.1 | XP_025003062.1 | 9 - 10 | 1777 | 69 | ||
| XM_025147295.1 | XP_025003063.1 | 8 - 9 | 1730 | 69 | ||
| XM_025147296.1 | XP_025003064.1 | 6 - 7 | 943 | 69 | ||
| GenBank mRNA | EF608155.1 | - | 8 - 9 | 1516 | 69 | |
More Information
Set Membership: |
|

CDH19
Genomic Map