Assay Details
| Gene Symbol: IGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101034.1 | NP_001094504.1 | 1 - 2 | 436 | 76 | |
| GenBank mRNA | EF624351.1 | - | 1 - 2 | 436 | 76 | |
More Information
Set Membership: |
|

IGFBP3
Genomic Map