Assay Details
| Gene Symbol: GCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101035.1 | NP_001094505.1 | 12 - 13 | 1228 | 60 | |
| XM_015295408.2 | XP_015150894.1 | 13 - 14 | 1439 | 60 | ||
| GenBank mRNA | EF624352.1 | - | 12 - 13 | 1228 | 60 | |
More Information
Set Membership: |
|

GCGR
Genomic Map