Assay Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101649.1 | NP_001095119.1 | 4 - 5 | 435 | 74 | |
| XM_015280787.2 | XP_015136273.1 | 4 - 5 | 3390 | 74 | ||
| XM_015280788.2 | XP_015136274.1 | 4 - 5 | 423 | 74 | ||
| XM_015280789.2 | XP_015136275.1 | 5 - 6 | 3511 | 74 | ||
| XM_015280790.2 | XP_015136276.1 | 4 - 5 | 401 | 74 | ||
| XM_015280791.2 | XP_015136277.1 | 5 - 6 | 3479 | 74 | ||
| XM_015280793.1 | XP_015136279.1 | 4 - 5 | 353 | 74 | ||
| XM_025144704.1 | XP_025000472.1 | 5 - 6 | 3528 | 74 | ||
| XM_025144705.1 | XP_025000473.1 | 5 - 6 | 3449 | 74 | ||
| GenBank mRNA | BG625023.1 | - | 4 - 5 | 414 | 74 | |
More Information
Set Membership: |
|

SNX24
Genomic Map