Assay Details
| Gene Symbol: NEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101730.1 | NP_001095200.1 | 1 - 2 | 1069 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEFM| Gene Symbol: NEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101730.1 | NP_001095200.1 | 1 - 2 | 1069 | 74 | |
Set Membership: |
|