Assay Details
| Gene Symbol: NOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101830.1 | NP_001095300.1 | 11 - 12 | 1557 | 76 | |
| XM_015278269.1 | XP_015133755.1 | 10 - 11 | 2625 | 76 | ||
| XM_015278270.2 | XP_015133756.1 | 11 - 12 | 2583 | 76 | ||
| XM_015278271.2 | XP_015133757.1 | 10 - 11 | 2487 | 76 | ||
| XM_015278272.1 | XP_015133758.1 | 11 - 12 | 2481 | 76 | ||
| XM_015278273.2 | XP_015133759.1 | 12 - 13 | 1600 | 76 | ||
More Information
Set Membership: |
|

NOX1
Genomic Map