Assay Details
| Gene Symbol: LOC771308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104958.1 | NP_001098428.1 | 3 - 4 | 291 | 70 | |
| XM_015299360.2 | XP_015154846.1 | 3 - 4 | 673 | 70 | ||
| XM_015299361.2 | XP_015154847.1 | 3 - 4 | 673 | 70 | ||
| GenBank mRNA | EF209054.1 | - | 3 - 4 | 580 | 70 | |
| EF581858.1 | - | 3 - 4 | 291 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015299360.2, XM_015299361.2 transcripts.
Set Membership: |
|

LOC771308
Genomic Map