Assay Details
| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105610.1 | NP_001099080.1 | 2 - 3 | 181 | 70 | |
| GenBank mRNA | BX933927.2 | - | 3 - 4 | 362 | 70 | |
| EU069394.1 | - | 2 - 3 | 181 | 70 | ||
More Information
Set Membership: |
|

RAMP2
Genomic Map