Assay Details
| Gene Symbol: CAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105664.2 | NP_001099134.1 | 2 - 3 | 260 | 63 | |
| GenBank mRNA | L01582.1 | - | 2 - 3 | 264 | 63 | |
More Information
Set Membership: |
|

CAV1
Genomic Map