Assay Details
| Gene Symbol: RAB23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110060.1 | NP_001103530.1 | 4 - 5 | 479 | 78 | |
| XM_015284828.2 | XP_015140314.1 | 5 - 6 | 78 | |||
| XM_015284829.2 | XP_015140315.1 | 5 - 6 | 78 | |||
| GenBank mRNA | BX929572.2 | - | 5 - 6 | 624 | 78 | |
| BX931809.1 | - | 5 - 6 | 641 | 78 | ||
| CR406263.1 | - | 5 - 6 | 629 | 78 | ||
| EU176872.1 | - | 4 - 5 | 479 | 78 | ||
More Information
Set Membership: |
|

RAB23
Genomic Map