Assay Details
| Gene Symbol: CALM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110364.1 | NP_001103834.1 | 3 - 4 | 238 | 128 | |
| XM_025150616.1 | XP_025006384.1 | 3 - 4 | 312 | 128 | ||
| GenBank mRNA | CN228694.1 | - | 3 - 4 | 239 | 128 | |
| CR387408.1 | - | 3 - 4 | 254 | 128 | ||
| DR430013.1 | - | 1 - 2 | 85 | 128 | ||
| M18355.1 | - | 1 - 2 | 142 | 128 | ||
More Information
Set Membership: |
|

CALM1
Genomic Map