Assay Details
| Gene Symbol: CNP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111259.1 | NP_001104729.1 | 1 - 2 | 98 | 61 | |
| GenBank mRNA | BX929576.2 | - | 1 - 2 | 130 | 61 | |
| CR405820.1 | - | 1 - 2 | 135 | 61 | ||
| DQ855411.1 | - | 1 - 2 | 98 | 61 | ||
More Information
Set Membership: |
|

CNP3
Genomic Map