Assay Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112809.1 | NP_001106280.1 | 3 - 4 | 288 | 77 | |
| GenBank mRNA | BU323301.1 | - | 3 - 4 | 77 | ||
| BX932407.2 | - | 3 - 4 | 271 | 77 | ||
| BX950842.1 | - | 3 - 4 | 77 | |||
| CV857320.1 | - | 3 - 4 | 77 | |||
More Information
Set Membership: |
|

HSBP1
Genomic Map