Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204114.4 | NP_989445.3 | 1 - 2 | 820 | 76 | |
| GenBank mRNA | AF125575.1 | - | 1 - 2 | 822 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIO2| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204114.4 | NP_989445.3 | 1 - 2 | 820 | 76 | |
| GenBank mRNA | AF125575.1 | - | 1 - 2 | 822 | 76 | |
Set Membership: |
|