Assay Details
| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205174.2 | NP_990505.1 | 6 - 7 | 2160 | 62 | |
| GenBank mRNA | S53899.1 | - | 6 - 7 | 2160 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKIL| Gene Symbol: SKIL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205174.2 | NP_990505.1 | 6 - 7 | 2160 | 62 | |
| GenBank mRNA | S53899.1 | - | 6 - 7 | 2160 | 62 | |
Set Membership: |
|