Assay Details
| Gene Symbol: CTCF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205332.5 | NP_990663.2 | 8 - 9 | 2195 | 67 | |
| XM_015279119.2 | XP_015134605.1 | 8 - 9 | 1866 | 67 | ||
| XM_015279120.2 | XP_015134606.1 | 8 - 9 | 1863 | 67 | ||
| XM_015279121.2 | XP_015134607.1 | 7 - 8 | 1724 | 67 | ||
| XM_025154151.1 | XP_025009919.1 | 8 - 9 | 1685 | 67 | ||
| GenBank mRNA | Z22605.1 | - | 7 - 8 | 2195 | 67 | |
More Information
Set Membership: |
|

CTCF
Genomic Map