Assay Details
| Gene Symbol: GABRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205345.2 | NP_990676.1 | 7 - 8 | 1595 | 67 | |
| XM_015293638.2 | XP_015149124.1 | 7 - 8 | 959 | 67 | ||
| GenBank mRNA | X54944.1 | - | 1595 | 67 | ||
More Information
Set Membership: |
|

GABRG2
Genomic Map