Assay Details
| Gene Symbol: ANP32E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006564.2 | NP_001006564.2 | 2 - 3 | 356 | 60 | |
| XM_015298326.2 | XP_015153812.1 | 2 - 3 | 427 | 60 | ||
| GenBank mRNA | AJ720498.1 | - | 2 - 3 | 392 | 60 | |
| AJ851397.1 | - | 2 - 3 | 356 | 60 | ||
More Information
Set Membership: |
|

ANP32E
Genomic Map