Assay Details
| Gene Symbol: UBLCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030613.3 | NP_001025784.2 | 3 - 4 | 339 | 56 | |
| XM_015293765.2 | XP_015149251.1 | 3 - 4 | 339 | 56 | ||
| XM_025154843.1 | XP_025010611.1 | 4 - 5 | 430 | 56 | ||
| XM_025154844.1 | XP_025010612.1 | 4 - 5 | 433 | 56 | ||
| XM_025154845.1 | XP_025010613.1 | 3 - 4 | 788 | 56 | ||
| GenBank mRNA | AJ720436.1 | - | 3 - 4 | 340 | 56 | |
More Information
Set Membership: |
|

UBLCP1
Genomic Map