Assay Details
| Gene Symbol: LPAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113710.1 | NP_001107182.1 | 1 - 2 | 737 | 89 | |
| XM_015290834.2 | XP_015146320.1 | 3 - 4 | 1622 | 89 | ||
| XM_015290835.2 | XP_015146321.1 | 3 - 4 | 1404 | 89 | ||
| XM_015290836.2 | XP_015146322.1 | 3 - 4 | 1617 | 89 | ||
| XM_025152900.1 | XP_025008668.1 | 3 - 4 | 2739 | 89 | ||
| XM_025152901.1 | XP_025008669.1 | 3 - 4 | 2734 | 89 | ||
| GenBank mRNA | EU339319.1 | - | 1 - 2 | 737 | 89 | |
More Information
Set Membership: |
|

LPAR3
Genomic Map