Assay Details
| Gene Symbol: FN3KRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114638.1 | NP_001108110.1 | 4 - 5 | 474 | 112 | |
| XM_015295139.2 | XP_015150625.1 | 4 - 5 | 473 | 112 | ||
| XM_025141584.1 | XP_024997352.1 | 4 - 5 | 535 | 112 | ||
More Information
Set Membership: |
|

FN3KRP
Genomic Map