Assay Details
| Gene Symbol: JMJD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114659.1 | NP_001108131.1 | 4 - 5 | 532 | 123 | |
| GenBank mRNA | BX929829.2 | - | 3 - 4 | 470 | 123 | |
| CR407278.1 | - | 3 - 4 | 328 | 123 | ||
More Information
Set Membership: |
|

JMJD7
Genomic Map