Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122763.1 | NP_001116235.1 | 3 - 4 | 272 | 75 | |
| GenBank mRNA | BU106104.1 | - | 2 - 3 | 249 | 75 | |
| BX934858.1 | - | 3 - 4 | 237 | 75 | ||
| CD765296.1 | - | 2 - 3 | 275 | 75 | ||
More Information
Set Membership: |
|

RPL37
Genomic Map