Assay Details
| Gene Symbol: AHR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204118.2 | NP_989449.2 | 9 - 10 | 1151 | 99 | |
| XM_015281254.2 | XP_015136740.1 | 6 - 7 | 832 | 99 | ||
| XM_025146991.1 | XP_025002759.1 | 9 - 10 | 1706 | 99 | ||
| GenBank mRNA | AF192502.2 | - | 9 - 10 | 1181 | 99 | |
| AF260832.1 | - | 9 - 10 | 1151 | 99 | ||
More Information
Set Membership: |
|

AHR1A
Genomic Map