Assay Details
| Gene Symbol: AHR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204118.2 | NP_989449.2 | 10 - 11 | 2427 | 140 | |
| XM_015281254.2 | XP_015136740.1 | 7 - 8 | 2108 | 140 | ||
| XM_025146991.1 | XP_025002759.1 | 10 - 11 | 2982 | 140 | ||
| GenBank mRNA | AF192502.2 | - | 10 - 11 | 2457 | 140 | |
| AF260832.1 | - | 10 - 11 | 2427 | 140 | ||
More Information
Set Membership: |
|

AHR1A
Genomic Map