Assay Details
| Gene Symbol: EDNRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204120.1 | NP_989451.1 | 8 - 9 | 1439 | 82 | |
| XM_015278144.2 | XP_015133630.1 | 7 - 8 | 1382 | 82 | ||
| XM_015278145.2 | XP_015133631.1 | 7 - 8 | 1382 | 82 | ||
| XM_015278146.2 | XP_015133632.1 | 7 - 8 | 1517 | 82 | ||
| XM_015278148.2 | XP_015133634.1 | 8 - 9 | 1458 | 82 | ||
| XM_015278149.2 | XP_015133635.1 | 8 - 9 | 1432 | 82 | ||
| XM_015278150.2 | XP_015133636.1 | 7 - 8 | 1374 | 82 | ||
| XM_015278151.2 | XP_015133637.1 | 8 - 9 | 1541 | 82 | ||
| GenBank mRNA | AB697059.1 | - | 6 - 7 | 1162 | 82 | |
| AB697060.1 | - | 6 - 7 | 1162 | 82 | ||
| AB697061.1 | - | 6 - 7 | 1162 | 82 | ||
| AB697062.1 | - | 6 - 7 | 1162 | 82 | ||
| AF472617.1 | - | 8 - 9 | 1439 | 82 | ||
More Information
Set Membership: |
|

EDNRB2
Genomic Map