Assay Details
| Gene Symbol: NBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204149.4 | NP_989480.2 | 2 - 3 | 422 | 87 | |
| GenBank mRNA | AB044337.1 | - | 2 - 3 | 385 | 87 | |
| AF421881.1 | - | 2 - 3 | 217 | 87 | ||
More Information
Set Membership: |
|

NBL1
Genomic Map