Assay Details
| Gene Symbol: MAPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204150.1 | NP_989481.1 | 4 - 5 | 634 | 71 | |
| XM_015275131.2 | XP_015130617.1 | 4 - 5 | 845 | 71 | ||
| GenBank mRNA | AY033635.1 | - | 4 - 5 | 634 | 71 | |
More Information
Set Membership: |
|

MAPK3
Genomic Map