Assay Details
| Gene Symbol: PSEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204163.2 | NP_989494.2 | 6 - 7 | 900 | 74 | |
| XM_015286913.2 | XP_015142399.1 | 7 - 8 | 1707 | 74 | ||
| XM_015286914.2 | XP_015142400.1 | 6 - 7 | 905 | 74 | ||
| GenBank mRNA | AY043492.1 | - | 6 - 7 | 854 | 74 | |
| BU202054.1 | - | 3 - 4 | 334 | 74 | ||
| DQ084362.1 | - | 5 - 6 | 769 | 74 | ||
More Information
Set Membership: |
|

PSEN1
Genomic Map